siRNA Details
Gene Symbol: Olfr1347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146385.1 | NP_666497.1 | 1 | 54 | ||
GenBank mRNA | BC128003.1 | - | 1 | 58 | 40135936 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
