siRNA Details
| Gene Symbol: Olfr1305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146401.2 | NP_666513.2 | 1 | 196 | ||
| GenBank mRNA | BC106975.1 | - | 1 | 189 | ||
| BC106976.1 | - | 1 | 189 | |||
| BC114640.1 | - | 1 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1305| Gene Symbol: Olfr1305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146401.2 | NP_666513.2 | 1 | 196 | ||
| GenBank mRNA | BC106975.1 | - | 1 | 189 | ||
| BC106976.1 | - | 1 | 189 | |||
| BC114640.1 | - | 1 | 195 | |||