siRNA Details
| Gene Symbol: Olfr1065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146408.2 | NP_666520.2 | 1 | 928 | ||
| GenBank mRNA | BC138401.1 | - | 1 | 928 | 8861421 | |
| BC138402.1 | - | 1 | 928 | 8861422 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1065| Gene Symbol: Olfr1065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146408.2 | NP_666520.2 | 1 | 928 | ||
| GenBank mRNA | BC138401.1 | - | 1 | 928 | 8861421 | |
| BC138402.1 | - | 1 | 928 | 8861422 | ||