siRNA Details
| Gene Symbol: Olfr462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146411.2 | NP_666523.2 | 1 | 909 | ||
| GenBank mRNA | BC108939.1 | - | 909 | 40041186 | ||
| BC108940.1 | - | 910 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr462| Gene Symbol: Olfr462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146411.2 | NP_666523.2 | 1 | 909 | ||
| GenBank mRNA | BC108939.1 | - | 909 | 40041186 | ||
| BC108940.1 | - | 910 | ||||