siRNA Details
| Gene Symbol: Olfr887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146423.1 | NP_666634.1 | 1 | 21 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr887| Gene Symbol: Olfr887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146423.1 | NP_666634.1 | 1 | 21 | ||