siRNA Details
| Gene Symbol: Olfr995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146434.1 | NP_666645.1 | 1 | 619 | ||
| GenBank mRNA | BC125413.1 | - | 1 | 700 | 40129928 | |
| BC125415.1 | - | 1 | 700 | 40129930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr995| Gene Symbol: Olfr995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146434.1 | NP_666645.1 | 1 | 619 | ||
| GenBank mRNA | BC125413.1 | - | 1 | 700 | 40129928 | |
| BC125415.1 | - | 1 | 700 | 40129930 | ||