siRNA Details
| Gene Symbol: Olfr1209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146461.2 | NP_666672.2 | 1 | 117 | ||
| GenBank mRNA | BC104269.1 | - | 1 | 117 | 40040821 | |
| BC104270.1 | - | 1 | 115 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1209| Gene Symbol: Olfr1209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146461.2 | NP_666672.2 | 1 | 117 | ||
| GenBank mRNA | BC104269.1 | - | 1 | 117 | 40040821 | |
| BC104270.1 | - | 1 | 115 | |||