siRNA Details
| Gene Symbol: Olfr91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182714.2 | NP_874373.2 | 1 | 637 | ||
| GenBank mRNA | BC108941.1 | - | 1 | 635 | ||
| BC108942.1 | - | 1 | 632 | |||
| BC119413.1 | - | 1 | 637 | 40042089 | ||
| BC119414.1 | - | 1 | 637 | 40042091 | ||

Olfr91
Genomic Map