siRNA Details
| Gene Symbol: Olfr899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146479.1 | NP_666690.1 | 1 | 29 | ||
| GenBank mRNA | BC104124.1 | - | 1 | 29 | 40044544 | |
| BC104125.1 | - | 1 | 29 | 40044545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr899| Gene Symbol: Olfr899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146479.1 | NP_666690.1 | 1 | 29 | ||
| GenBank mRNA | BC104124.1 | - | 1 | 29 | 40044544 | |
| BC104125.1 | - | 1 | 29 | 40044545 | ||