siRNA Details
| Gene Symbol: Olfr730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146493.2 | NP_666704.2 | 1 | 60 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr730| Gene Symbol: Olfr730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146493.2 | NP_666704.2 | 1 | 60 | ||