siRNA Details
| Gene Symbol: Olfr484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146499.1 | NP_666710.1 | 1 | 648 | ||
| GenBank mRNA | BC137823.1 | - | 1 | 683 | 8860842 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr484| Gene Symbol: Olfr484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146499.1 | NP_666710.1 | 1 | 648 | ||
| GenBank mRNA | BC137823.1 | - | 1 | 683 | 8860842 | |