siRNA Details
| Gene Symbol: Olfr319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146500.2 | NP_666711.2 | 1 | 325 | ||
| GenBank mRNA | BC104265.1 | - | 1 | 325 | 40040340 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr319| Gene Symbol: Olfr319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146500.2 | NP_666711.2 | 1 | 325 | ||
| GenBank mRNA | BC104265.1 | - | 1 | 325 | 40040340 | |