siRNA Details
| Gene Symbol: Olfr279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001807.1 | NP_001001807.1 | 1 | 889 | ||
| GenBank mRNA | BC106968.1 | - | 1 | 889 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr279| Gene Symbol: Olfr279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001807.1 | NP_001001807.1 | 1 | 889 | ||
| GenBank mRNA | BC106968.1 | - | 1 | 889 | ||