siRNA Details
| Gene Symbol: Olfr1364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146540.2 | NP_666751.2 | 1 | 89 | ||
| GenBank mRNA | BC127986.1 | - | 1 | 93 | 40135567 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1364| Gene Symbol: Olfr1364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146540.2 | NP_666751.2 | 1 | 89 | ||
| GenBank mRNA | BC127986.1 | - | 1 | 93 | 40135567 | |