siRNA Details
| Gene Symbol: Olfr810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146550.1 | NP_666761.1 | 1 | 604 | ||
| GenBank mRNA | BC120749.1 | - | 1 | 631 | 40129672 | |
| BC120751.1 | - | 1 | 631 | 40129674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr810| Gene Symbol: Olfr810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146550.1 | NP_666761.1 | 1 | 604 | ||
| GenBank mRNA | BC120749.1 | - | 1 | 631 | 40129672 | |
| BC120751.1 | - | 1 | 631 | 40129674 | ||