siRNA Details
| Gene Symbol: Olfr836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146564.2 | NP_666775.1 | 1 | 317 | ||
| GenBank mRNA | BC106988.1 | - | 1 | 317 | 40043348 | |
| BC106989.1 | - | 1 | 317 | 40043350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr836| Gene Symbol: Olfr836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146564.2 | NP_666775.1 | 1 | 317 | ||
| GenBank mRNA | BC106988.1 | - | 1 | 317 | 40043348 | |
| BC106989.1 | - | 1 | 317 | 40043350 | ||