siRNA Details
| Gene Symbol: Olfr837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146565.2 | NP_666776.1 | 1 | 377 | ||
| GenBank mRNA | BC140981.1 | - | 1 | 377 | 9055593 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr837| Gene Symbol: Olfr837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146565.2 | NP_666776.1 | 1 | 377 | ||
| GenBank mRNA | BC140981.1 | - | 1 | 377 | 9055593 | |