siRNA Details
| Gene Symbol: Olfr828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146605.1 | NP_666816.1 | 1 | 218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr828| Gene Symbol: Olfr828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146605.1 | NP_666816.1 | 1 | 218 | ||