siRNA Details
| Gene Symbol: Olfr1158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146645.2 | NP_666856.2 | 1 | 427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1158| Gene Symbol: Olfr1158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146645.2 | NP_666856.2 | 1 | 427 | ||