siRNA Details
| Gene Symbol: Olfr444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146656.1 | NP_666867.1 | 1 | 471 | ||
| GenBank mRNA | BC145664.1 | - | 1 | 512 | 40130967 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr444| Gene Symbol: Olfr444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146656.1 | NP_666867.1 | 1 | 471 | ||
| GenBank mRNA | BC145664.1 | - | 1 | 512 | 40130967 | |