siRNA Details
| Gene Symbol: Olfr1136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146659.3 | NP_666870.2 | 1 | 313 | ||
| GenBank mRNA | BC120618.1 | - | 1 | 315 | 40129541 | |
| BC120620.1 | - | 1 | 315 | 40129543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1136| Gene Symbol: Olfr1136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146659.3 | NP_666870.2 | 1 | 313 | ||
| GenBank mRNA | BC120618.1 | - | 1 | 315 | 40129541 | |
| BC120620.1 | - | 1 | 315 | 40129543 | ||