siRNA Details
| Gene Symbol: Olfr1442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146697.2 | NP_666908.2 | 1 | 346 | ||
| GenBank mRNA | BC119095.1 | - | 1 | 346 | 8733844 | |
| BC119097.1 | - | 1 | 346 | 8733846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1442| Gene Symbol: Olfr1442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146697.2 | NP_666908.2 | 1 | 346 | ||
| GenBank mRNA | BC119095.1 | - | 1 | 346 | 8733844 | |
| BC119097.1 | - | 1 | 346 | 8733846 | ||