siRNA Details
Gene Symbol: Olfr1442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146697.2 | NP_666908.2 | 1 | 74 | ||
GenBank mRNA | BC119095.1 | - | 1 | 74 | 8733844 | |
BC119097.1 | - | 1 | 74 | 8733846 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146697.2 | NP_666908.2 | 1 | 74 | ||
GenBank mRNA | BC119095.1 | - | 1 | 74 | 8733844 | |
BC119097.1 | - | 1 | 74 | 8733846 |