siRNA Details
| Gene Symbol: Olfr430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146718.2 | NP_666929.2 | 1 | 905 | ||
| GenBank mRNA | BC106773.1 | - | 1 | 850 | ||
| BC106774.1 | - | 1 | 847 | |||
| BC126860.1 | - | 1 | 844 | |||
| BC126861.1 | - | 1 | 844 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr430| Gene Symbol: Olfr430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146718.2 | NP_666929.2 | 1 | 905 | ||
| GenBank mRNA | BC106773.1 | - | 1 | 850 | ||
| BC106774.1 | - | 1 | 847 | |||
| BC126860.1 | - | 1 | 844 | |||
| BC126861.1 | - | 1 | 844 | |||