siRNA Details
| Gene Symbol: Olfr483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146735.1 | NP_666946.1 | 1 | 329 | ||
| GenBank mRNA | BC116910.1 | - | 1 | 361 | 40126229 | |
| BC116912.1 | - | 1 | 361 | 40126231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr483| Gene Symbol: Olfr483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146735.1 | NP_666946.1 | 1 | 329 | ||
| GenBank mRNA | BC116910.1 | - | 1 | 361 | 40126229 | |
| BC116912.1 | - | 1 | 361 | 40126231 | ||