siRNA Details
| Gene Symbol: Olfr494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146737.1 | NP_666948.1 | 1 | 596 | ||
| GenBank mRNA | BC127987.1 | - | 600 | 40135591 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr494| Gene Symbol: Olfr494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146737.1 | NP_666948.1 | 1 | 596 | ||
| GenBank mRNA | BC127987.1 | - | 600 | 40135591 | ||