siRNA Details
| Gene Symbol: Olfr502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146739.1 | NP_666950.1 | 1 | 601 | ||
| GenBank mRNA | BC119235.1 | - | 1 | 620 | 8733984 | |
| BC120678.1 | - | 1 | 620 | 40129601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr502| Gene Symbol: Olfr502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146739.1 | NP_666950.1 | 1 | 601 | ||
| GenBank mRNA | BC119235.1 | - | 1 | 620 | 8733984 | |
| BC120678.1 | - | 1 | 620 | 40129601 | ||