siRNA Details
Gene Symbol: Olfr689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146750.1 | NP_666961.1 | 1 | 945 | ||
GenBank mRNA | BC119379.1 | - | 1 | 969 | 8734128 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146750.1 | NP_666961.1 | 1 | 945 | ||
GenBank mRNA | BC119379.1 | - | 1 | 969 | 8734128 |