siRNA Details
| Gene Symbol: Olfr1245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146788.2 | NP_666999.2 | 1 | 239 | ||
| GenBank mRNA | BC138384.1 | - | 1 | 239 | 8861404 | |
| BC138385.1 | - | 1 | 239 | 8861405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1245| Gene Symbol: Olfr1245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146788.2 | NP_666999.2 | 1 | 239 | ||
| GenBank mRNA | BC138384.1 | - | 1 | 239 | 8861404 | |
| BC138385.1 | - | 1 | 239 | 8861405 | ||