siRNA Details
| Gene Symbol: Olfr983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146827.2 | NP_667038.1 | 2 | 724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr983| Gene Symbol: Olfr983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146827.2 | NP_667038.1 | 2 | 724 | ||