siRNA Details
| Gene Symbol: Olfr985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146855.2 | NP_667066.1 | 1 | 233 | ||
| GenBank mRNA | BC138430.1 | - | 1 | 233 | 8861450 | |
| BC138431.1 | - | 1 | 233 | 8861451 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr985| Gene Symbol: Olfr985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146855.2 | NP_667066.1 | 1 | 233 | ||
| GenBank mRNA | BC138430.1 | - | 1 | 233 | 8861450 | |
| BC138431.1 | - | 1 | 233 | 8861451 | ||