siRNA Details
| Gene Symbol: Olfr898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146871.2 | NP_667082.2 | 1 | 72 | ||
| GenBank mRNA | BC127995.1 | - | 1 | 76 | 40135758 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr898| Gene Symbol: Olfr898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146871.2 | NP_667082.2 | 1 | 72 | ||
| GenBank mRNA | BC127995.1 | - | 1 | 76 | 40135758 | |