siRNA Details
| Gene Symbol: Olfr895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146875.2 | NP_667086.2 | 1 | 438 | ||
| GenBank mRNA | BC127996.1 | - | 1 | 442 | ||
| BC152319.1 | - | 1 | 442 | 40135771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr895| Gene Symbol: Olfr895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146875.2 | NP_667086.2 | 1 | 438 | ||
| GenBank mRNA | BC127996.1 | - | 1 | 442 | ||
| BC152319.1 | - | 1 | 442 | 40135771 | ||