siRNA Details
| Gene Symbol: Olfr876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146883.2 | NP_667094.2 | 1 | 606 | ||
| GenBank mRNA | BC120726.1 | - | 1 | 606 | 40129649 | |
| BC120732.1 | - | 1 | 606 | 40129655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr876| Gene Symbol: Olfr876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146883.2 | NP_667094.2 | 1 | 606 | ||
| GenBank mRNA | BC120726.1 | - | 1 | 606 | 40129649 | |
| BC120732.1 | - | 1 | 606 | 40129655 | ||