siRNA Details
| Gene Symbol: Olfr1299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146884.2 | NP_667095.2 | 2 | 562 | ||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Rat: NM_001000372.
					            	This siRNA also targets the following transcripts in Rat: NM_001000372.Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Olfr1299
Olfr1299 Genomic Map
Genomic Map Back To Top
 Back To Top