siRNA Details
Gene Symbol: Olfr1348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146913.1 | NP_667124.1 | 1 | 33 | ||
GenBank mRNA | BC120570.1 | - | 1 | 129 | 40129493 | |
BC137836.1 | - | 1 | 129 | 8860855 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
