siRNA Details
| Gene Symbol: Olfr347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146943.1 | NP_667154.1 | 1 | 795 | ||
| GenBank mRNA | BC127977.1 | - | 1 | 799 | 40135187 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr347| Gene Symbol: Olfr347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146943.1 | NP_667154.1 | 1 | 795 | ||
| GenBank mRNA | BC127977.1 | - | 1 | 799 | 40135187 | |