siRNA Details
| Gene Symbol: Olfr1233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146972.1 | NP_667183.1 | 1 | 561 | ||
| GenBank mRNA | BC119149.1 | - | 1 | 609 | 8733898 | |
| BC119151.1 | - | 1 | 609 | 8733900 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1233| Gene Symbol: Olfr1233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146972.1 | NP_667183.1 | 1 | 561 | ||
| GenBank mRNA | BC119149.1 | - | 1 | 609 | 8733898 | |
| BC119151.1 | - | 1 | 609 | 8733900 | ||