siRNA Details
| Gene Symbol: Olfr206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146991.1 | NP_667202.1 | 1 | 211 | ||
| GenBank mRNA | BC138396.1 | - | 1 | 238 | 8861416 | |
| BC138397.1 | - | 1 | 238 | 8861417 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr206| Gene Symbol: Olfr206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146991.1 | NP_667202.1 | 1 | 211 | ||
| GenBank mRNA | BC138396.1 | - | 1 | 238 | 8861416 | |
| BC138397.1 | - | 1 | 238 | 8861417 | ||