siRNA Details
| Gene Symbol: Olfr178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146997.2 | NP_667208.2 | 1 | 905 | ||
| GenBank mRNA | BC111099.1 | - | 1 | 904 | ||
| BC127221.1 | - | 1 | 903 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr178| Gene Symbol: Olfr178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146997.2 | NP_667208.2 | 1 | 905 | ||
| GenBank mRNA | BC111099.1 | - | 1 | 904 | ||
| BC127221.1 | - | 1 | 903 | |||