siRNA Details
Gene Symbol: Olfr1413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147037.2 | NP_667248.1 | 1 | 492 | ||
GenBank mRNA | BC116898.1 | - | 1 | 492 | 40126217 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147037.2 | NP_667248.1 | 1 | 492 | ||
GenBank mRNA | BC116898.1 | - | 1 | 492 | 40126217 |