siRNA Details
| Gene Symbol: Olfr683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147045.1 | NP_667256.1 | 1 | 945 | ||
| GenBank mRNA | BC111830.1 | - | 1 | 938 | ||
| BC148203.1 | - | 1 | 945 | 40055104 | ||
More Information
Set Membership: |
|

Olfr683
Genomic Map