siRNA Details
Gene Symbol: Olfr569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147088.1 | NP_667299.1 | 1 | 920 | ||
GenBank mRNA | BC126864.1 | - | 1 | 920 | 40042577 | |
BC126865.1 | - | 1 | 920 | 40042578 | ||
BC127048.1 | - | 1 | 920 | 40056696 |
