siRNA Details
| Gene Symbol: Ehd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153068.3 | NP_694708.2 | 2 | 535 | ||
| GenBank mRNA | AK029240.1 | - | 2 | 479 | ||
| AK031203.1 | - | 2 | 537 | |||
| AK046566.1 | - | 2 | 477 | |||
| AK048356.1 | - | 2 | 480 | |||
| AK050243.1 | - | 2 | 515 | |||
| AK084455.1 | - | 2 | 478 | |||
| AK085016.1 | - | 2 | 478 | |||
| AY531389.1 | - | 2 | 474 | |||
| BC027084.1 | - | 3 | 182 | 5356224 | ||
| BC031731.1 | - | 400 | ||||
| BC113161.1 | - | 3 | 493 | 40060184 | ||

Ehd2
Genomic Map