siRNA Details
| Gene Symbol: Ehd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153068.3 | NP_694708.2 | 4 | 755 | ||
| GenBank mRNA | AK029240.1 | - | 4 | 699 | ||
| AK031203.1 | - | 4 | 757 | |||
| AK046566.1 | - | 4 | 697 | |||
| AK048356.1 | - | 4 | 700 | |||
| AK050243.1 | - | 4 | 735 | |||
| AK084455.1 | - | 4 | 698 | |||
| AK085016.1 | - | 4 | 698 | |||
| AY531389.1 | - | 4 | 694 | |||
| BC027084.1 | - | 5 | 402 | 5356224 | ||
| BC031731.1 | - | 620 | ||||
| BC113161.1 | - | 5 | 713 | 40060184 | ||

Ehd2
Genomic Map