siRNA Details
| Gene Symbol: Otos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153114.2 | NP_694754.1 | 5 | 466 | ||
| GenBank mRNA | AK083090.1 | - | 4 | 2754 | ||
| AY078071.1 | - | 4 | 214 | |||
| BC100323.1 | - | 5 | 466 | 30788363 | ||
| BC132494.1 | - | 4 | 385 | 40130771 | ||
| BC132498.1 | - | 4 | 385 | 40130775 | ||

Otos
Genomic Map