siRNA Details
| Gene Symbol: Gga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252067.1 | NP_001238996.1 | 12 | 1297 | ||
| NM_173048.3 | NP_766636.2 | 13 | 1531 | |||
| GenBank mRNA | AK031086.1 | - | 13 | 1503 | ||
| AK082327.1 | - | 12 | 1261 | |||
| AK122212.2 | - | 13 | 1500 | |||
| AK132183.1 | - | 13 | 1499 | |||
| AK147882.1 | - | 13 | 1499 | |||
| BC024266.1 | - | 2138 | ||||
| BC056424.1 | - | 13 | 1485 | |||
| BC085617.1 | - | 10 | 3296 | |||
| BC110676.1 | - | 11 | 2014 | |||

Gga3
Genomic Map