siRNA Details
| Gene Symbol: Avpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011924.2 | NP_036054.1 | 1 | 1270 | ||
| GenBank mRNA | AK033416.1 | - | 1 | 304 | ||
| AK050523.1 | - | 1 | 352 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Avpr1b| Gene Symbol: Avpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011924.2 | NP_036054.1 | 1 | 1270 | ||
| GenBank mRNA | AK033416.1 | - | 1 | 304 | ||
| AK050523.1 | - | 1 | 352 | |||
Set Membership: |
|