siRNA Details
| Gene Symbol: Clcn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011929.2 | NP_036059.1 | 22 | 2425 | ||
| GenBank mRNA | AK165362.1 | - | 22 | 2475 | ||
| AK179167.1 | - | 2 | 69 | |||
| AK197450.1 | - | 2 | 69 | |||
| AK220319.1 | - | 16 | 1915 | |||

Clcn6
Genomic Map