siRNA Details
| Gene Symbol: Clcn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011930.3 | NP_036060.1 | 19 | 1769 | ||
| GenBank mRNA | AK030444.1 | - | 19 | 1770 | ||
| AK032674.1 | - | 6 | 499 | |||
| AK128981.1 | - | 18 | 1695 | |||
| AK150395.1 | - | 19 | 1680 | |||
| BC050907.1 | - | 19 | 1741 | 5698121 | ||
| BC053049.1 | - | 19 | 1741 | 5698121 | ||
| BC054799.1 | - | 19 | 1749 | 6849205 | ||

Clcn7
Genomic Map